Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgA Antibody (BFG-267), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142270AF350
Description
Human IgA Monoclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Human IgA | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Human | |
| IgG1 |
| ELISA, Immunohistochemistry | |
| BFG-267 | |
| ELISA, Immunohistochemistry | |
| CD79A;IGA;Igalpha;Ig-alpha;Ly54;Ly-54;MB1;MB-1 | |
| Purified alpha-chain (Fc) of human IgA | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction