Learn More
Human IgA Antibody (BFG-267), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP142270G
Catalog No. NBP142270G
Description
Description
Human IgA Monoclonal specifically detects Human IgA in Human samples. It is validated for ELISA.
Specifications
Specifications
Human IgA
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified alpha-chain (Fc) of human IgA
0.2 mL
Primary
Human
Purified
ELISA
BFG-267
ELISA
Mouse
Ion exchange chromatography
RUO
This monoclonal antibody reacts with human IgA, alpha-chain. No cross reaction with human plasma proteins other then human IgA. No cross reaction with purified human IgE, IgG and IgM. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.