Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgA Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $538.00
Specifications
Antigen | IgA |
---|---|
Clone | 2217A |
Dilution | Western Blot 0.2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4787SP
![]() |
R&D Systems
MAB4787SP |
25 μg |
Each for $227.50
|
|
|||||
MAB4787100
![]() |
R&D Systems
MAB4787100 |
100 μg |
Each for $538.00
|
|
|||||
Description
IgA Monoclonal specifically detects IgA in Human samples. It is validated for Western Blot.Specifications
IgA | |
Western Blot 0.2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
3493 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2217A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Immunoglobulin A | |
Purified IgA from human serum | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IgA in Western blots. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title