Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgA Isotype Control, Novus Biologicals™
SDP

Catalog No. DDXCH06P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXCH06P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXCH06P100 Supplier Novus Biologicals Supplier No. DDXCH06P100
Add to Cart
Add to Cart

Human Polyclonal Antibody

IgA Polyclonal specifically detects IgA in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen IgA
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500
Formulation Tris-NaCl, pH 8. with No Preservative
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Human
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3493
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgA
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.