Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgA Monoclonal Antibody (KT40)
GREENER_CHOICE

Catalog No. PIMA549595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549595 Supplier Invitrogen™ Supplier No. MA549595
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product was used in an Indirect ELISA.

Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgA
Applications ELISA
Classification Monoclonal
Clone KT40
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IGHA1
Gene Accession No. P01876, P01877
Gene Alias A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)
Gene Symbols IGHA1, IGHA2
Host Species Mouse
Immunogen Recombinant Human IgA2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3493, 3494
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.