Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgA Recombinant Rabbit Monoclonal Antibody (10Q6M3)
GREENER_CHOICE

Catalog No. PIMA542727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542727 Supplier Invitrogen™ Supplier No. MA542727
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Human plasma. Immunogen sequence: ASPTSPKVFP LSLCSTQPDG NVVIACLVQG FFPQEPLSVT WSESGQGVTA RNFPPSQDAS GDLYTTSSQL TLPATQCLAG KSVTCHVKHY TNPSQDVTVP CPVPSTPPTP SPSTPPTPSP SCCHPRLSLH RPALEDLLLG SEANLTCTLT GLRDASGVTF TWTPSSGKSA VQGPPERDLC GCYSVSSVLP GCAEPWNHGK TFTCTAAYPE SKTPLTATLS KSGNTFRPEV HLLPPPSEEL ALNELVTLTC LARGFSPKDV LVRWLQGSQE LPREKYLTWA SRQEPSQGTT TFAVTSILRV AAEDWKKGDT FSCMVGHEAL PLAFTQKTID RLAGKPTHVN VSVVMAEVDG TCY.

Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgA
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 10Q6M3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IGHA1
Gene Accession No. P01876, P01877
Gene Alias A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)
Gene Symbols IGHA1, IGHA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-353 of human IgA (P01876).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3493, 3494
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.