Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgA2 Rabbit anti-Human, Clone: RM125 (Biotin), Abnova™

Catalog No. 89035305 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-305 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-305 Supplier Abnova Corporation Supplier No. MAB14953
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against native human IgA2.

  • High specificity
  • High reactivity

Specifications

Antigen Human IgA2
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone RM125
Conjugate Biotin
Description Rabbit monoclonal antibody raised against native human IgA2.
Dilution ELISA (0.05-0.2 ug/well for capture, 0.05-0.2 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-1 ug/mL) The optimal working dilution should be deter
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P01877
Gene Symbols IGHA2
Host Species Rabbit
Immunogen Native purified human IgA2.
Purification Method Protein A purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3494
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.