Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgD Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9857T100UG
This item is not returnable.
View return policy
Description
IgD Monoclonal specifically detects IgD in Human samples. It is validated for Flow Cytometry.Specifications
IgD | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human IgD in Flow Cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
IA6-2 | |
Flow Cytometry 0.25-1 μg/106 cells | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Human IgD, | |
100 μg | |
3495 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction