Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGDCC3 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8559V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8559V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8559V100 Supplier R&D Systems Supplier No. FAB8559V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGDCC3 Monoclonal specifically detects IGDCC3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGDCC3
Applications Flow Cytometry
Classification Monoclonal
Clone 920038
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q81VU1
Gene Alias HsT18880, immunoglobulin superfamily DCC subclass member 3, immunoglobulin superfamily, DCC subclass, member 3, PUNC, Putative neuronal cell adhesion moleculePUNCHsT18880
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IGDCC3 Met1-Gly640 Accession # Q81VU1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9543
Test Specificity Detects human IGDCC3 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.