Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. G107C
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G107C 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. G107C Supplier R&D Systems Supplier No. G107C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgE Polyclonal specifically detects IgE in Human samples. It is validated for .

Specifications

Antigen IgE
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Hidden
Gene Alias IgE, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Goat
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3497
Test Specificity This antibody reacts with heavy chains on IgE but not with other immunoglobulins in human serum.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Store the unopened product at 2 - 8degrees C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.