Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgE Rabbit anti-Human, Clone: RM122 (Biotin), Abnova™

Catalog No. 89035307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-307 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-307 Supplier Abnova Corporation Supplier No. MAB14955
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against native human IgE.

  • High specificity
  • High reactivity

Specifications

Antigen Human IgE
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RM122
Conjugate Biotin
Description Rabbit monoclonal antibody raised against native human IgE.
Dilution ELISA (0.01-0.1 ug/well for capture, 0.01-0.1 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P01854
Gene Alias IgE
Gene Symbols IGHE
Host Species Rabbit
Immunogen Native purified human IgE.
Purification Method Protein A purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3497
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.