Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGF-I/IGF-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$214.00 - $2020.00
Specifications
Antigen | IGF-I/IGF-1 |
---|---|
Clone | 56408 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGF-I/IGF-1 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P01343 | |
3479 | |
E. coli-derived recombinant human IGF-I/IGF-1 Gly49-Ala118 Accession # P01343 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IGF-I/IGF-1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) IGF-I/IGF-1, rmIGF-II/IGF-2, or recombinant human IGF-II/IGF-2. |
56408 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C | |
IGF1 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title