Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGF-I/IGF-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | IGF-I/IGF-1 |
---|---|
Clone | 997211 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2913SP
![]() |
R&D Systems
MAB2913SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2913100
![]() |
R&D Systems
MAB2913100 |
100 μg |
Each for $467.50
|
|
|||||
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human samples. It is validated for Immunocytochemistry.Specifications
IGF-I/IGF-1 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Lyophilized | |
Human | |
P05019 | |
3479 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
997211 | |
Immunocytochemistry | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C | |
E. coli-derived recombinant human IGF-I/IGF-1, Gly49-Ala118, Accession # P05019 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IGF-I/IGF-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title