Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGF-I R/IGF1R PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB391P100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB391P100 100 Tests
FAB391P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB391P100 Supplier R&D Systems Supplier No. FAB391P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGF-I R
Applications Flow Cytometry
Classification Monoclonal
Clone 33255
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P08069
Gene Alias CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IGF-I R Glu31-Asn932 Accession # P08069
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Test Specificity Detects human IGF-I R in sandwich ELISAs and Western blots. In sandwich ELISAs, less than 0.15% cross-reactivity or interference was observed with recombinant human (rh) IGF-I, rhIGF-II, rhIL-3 R alpha , rhIL-9 R, and rhTGF-beta RII.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.