Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGF-II/IGF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB292100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB292100 100 μg
MAB292SP 25 μg
MAB292500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB292100 Supplier R&D Systems Supplier No. MAB292100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Neutralization.

Specifications

Antigen IGF-II
Applications Neutralization
Classification Monoclonal
Clone 75015
Conjugate Unconjugated
Dilution Neutralization 2-12 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01344.1
Gene Alias C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A
Gene Symbols IGF2
Host Species Mouse
Immunogen E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3481
Test Specificity Detects human IGF-II in direct ELISA and Western blots. This antibody shows 100% cross-reactivity with recombinant mouse (rm) IGF-II and no cross-reactivity with recombinant human IGF-I or rmIGF-I.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.