Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGF-II/IGF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $1973.00
Specifications
Antigen | IGF-II |
---|---|
Clone | 75014 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2921SP
![]() |
R&D Systems
MAB2921SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2921100
![]() |
R&D Systems
MAB2921100 |
100 μg |
Each for $538.00
|
|
|||||
MAB2921500
![]() |
R&D Systems
MAB2921500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot.Specifications
IGF-II | |
Western Blot 2 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P01344.1 | |
3481 | |
E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IGF-II in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10 - 50% cross-reactivity with recombinant mouse IGF-II. |
75014 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
IGF2 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title