Learn More
R&D Systems™ Human IGF-II/IGF2 Quantikine ELISA Kit

Description
Includes
E. coli-expressed recombinant human IGF-II
Specifications
Specifications
| Description | C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PP9974, somatomedin-A |
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Human |
| Assay Sensitivity | 5.41 pg/mL |
| Assay Range | 12.50 to 800 pg/mL |
| Cross Reactivity | <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3481 |
| Sample Type | Serum-free Cell Culture Supernates (25uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Urine (25uL) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.