Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGF-II R/IGF2R Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2447
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2447 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2447 Supplier R&D Systems Supplier No. BAF2447
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IGF-II R/IGF2R Polyclonal specifically detects IGF-II R/IGF2R in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen IGF-II R/Mannose 6 Phosphate Receptor (Cation independent)
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P11717
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF2R, IGF-II receptor, IGFIIR, IGF-IIR, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRI, MPRIM6PR
Gene Symbols IGF2R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGF-II R Ser1510-Phe2108 Accession # P11717
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity Detects human IGF-II R in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human (rh) IGF-I R, rhIGF-I, and rhIGF-II is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.