Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB674SP
This item is not returnable.
View return policy
Description
IGFBP-2 Monoclonal specifically detects IGFBP-2 in Human samples. It is validated for Western Blot.Specifications
IGFBP-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3485 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
92320 | |
Western Blot 1 ug/mL | |
CAA34373 | |
IGFBP2 | |
Mouse myeloma cell line NS0-derived recombinant human IGFBP-2 Glu40-Gln328 Accession # CAA34373 | |
25 μg | |
Primary | |
Detects human IGFBP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IGFBP-1, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, rhIGFBP-6, recombinant mouse (rm) IGFBP-3, rmIGFBP-5, or rmIGFBP-6 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction