Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$214.00 - $2020.00
Specifications
Antigen | IGFBP-3 |
---|---|
Clone | 84728 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IGFBP-3 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
IGFBP3 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
84728 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Mouse | |
Human | |
CAA46087 | |
3486 | |
Mouse myeloma cell line NS0-derived recombinant human IGFBP-3 Gly28-Lys291 Accession # CAA46087 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IGFBP-3 in ELISAs and Western blots. In sandwich immunoassays, less than 0.25% cross-reactivity with recombinant human (rh) IGF-I R, rhIGFBP-1, rhIGFBP-2, rhIGFBP-4, rhIGFBP-5, and rhIGFBP-6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title