Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$188.28 - $579.52
Specifications
| Antigen | IGFBP-5 |
|---|---|
| Clone | 145619 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8751SP
![]() |
R&D Systems
MAB8751SP |
25 μg |
Each for $188.28
|
|
|||||
MAB8751100
![]() |
R&D Systems
MAB8751100 |
100 μg |
Each for $579.52
|
|
|||||
MAB8751500
![]() |
R&D Systems
MAB8751500 |
500 μg | N/A | N/A | N/A | ||||
Description
IGFBP-5 Monoclonal specifically detects IGFBP-5 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IGFBP-5 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IBP5IBP-5, IGF-binding protein 5, IGFBP5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
| IGFBP5 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 145619 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P24593 | |
| 3488 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-5 Glu28-Glu272 Accession # P24593 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGFBP-5 in ELISAs and Western blots. In ELISAs, this antibody shows 5% cross-reactivity with recombinant mouse (rm) IGFBP-5 and no cross-reactivity with rhIGFBP-2, -3, -4, -6, rmIGFBP-1, -2, -3, -6, rhIGFBP-rp1, rmIGF-I, rmIGF-II, rhIGF-I, or rhIGF-II. In Western blots, this antibody does not cross-react with rhIGFBP-1, -2, -3, -4, -6, or -7. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title