Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGFBP-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8761
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8761 500 μg
MAB8761SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8761 Supplier R&D Systems Supplier No. MAB8761
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

IGFBP-6 Monoclonal specifically detects IGFBP-6 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IGFBP-6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 110211
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P24592
Gene Alias IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6
Gene Symbols IGFBP6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGFBP-6 Ala25-Gly240 Accession # P24592
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3489
Test Specificity Detects human IGFBP-6 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IGFBP-1, -2, -3, -4, -5, rmIGFBP-3, -5, or -6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.