Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1334500
This item is not returnable.
View return policy
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Western Blot, ELISA.Specifications
IGFBP-rp1/IGFBP-7 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, ELISA | |
AAA16187 | |
IGFBP7 | |
Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1/IGFBP-7 Asp30-Arg277 Accession # AAA16187 | |
500 μg | |
Primary | |
Detects human IGFBP-rp1/IGFBP-7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IGFBP-1, -2, -3, -4, -5, or -6 is observed. | |
Human | |
Supernatant |
Western Blot, ELISA | |
192520 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3490 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction