Learn More
Human IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 6 publications
Supplier:  R&D Systems AF1334
Catalog No. AF1334
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
IGFBP-rp1/IGFBP-7 Polyclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
IGFBP-rp1/IGFBP-7
Polyclonal
Unconjugated
AAA16187
IGFBP7
Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1 Arg98-Arg277 Accession # AAA16187
100 μg
Primary
Detects human IGFBP-rp1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IGFBP-L1, rhIGFBP-rp10 and recombinant mouse IGFBP-7 is observed.
Human
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor
Goat
Affinity Purified
RUO
3490
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.