Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGFBP-rP10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5137
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5137 Supplier R&D Systems Supplier No. AF5137
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

IGFBP-rP10/KAZALD1 Polyclonal specifically detects IGFBP-rP10/KAZALD1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IGFBP-rP10/KAZALD1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q96I82
Gene Alias bone- and odontoblast-expressed gene 1, BONO1, FKSG28, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBPrP10, IGFBP-rP10, KAZALD1, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184
Gene Symbols KAZALD1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGFBP-rP10 isoform 1 Arg31-Tyr304 Accession # Q96I82
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81621
Test Specificity Detects human IGFBP-rP10 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse IGFBP-rP10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.