Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-rP10 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF5137
This item is not returnable.
View return policy
Description
IGFBP-rP10/KAZALD1 Polyclonal specifically detects IGFBP-rP10/KAZALD1 in Human samples. It is validated for Western Blot.Specifications
IGFBP-rP10/KAZALD1 | |
Polyclonal | |
Biotin | |
Q96I82 | |
KAZALD1 | |
Mouse myeloma cell line NS0-derived recombinant human IGFBP-rP10 isoform 1 Arg31-Tyr304 Accession # Q96I82 | |
50 μg | |
Primary | |
Detects human IGFBP-rP10 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse IGFBP-rP10 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot | |
bone- and odontoblast-expressed gene 1, BONO1, FKSG28, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBPrP10, IGFBP-rP10, KAZALD1, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184 | |
Sheep | |
Affinity Purified | |
RUO | |
81621 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction