Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGFL-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5970SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5970SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5970SP Supplier R&D Systems Supplier No. MAB5970SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGFL-3 Monoclonal specifically detects IGFL-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IGFL-3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 536825
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6UXB1
Gene Alias IGFL3, UNQ483
Gene Symbols IGFL3
Host Species Mouse
Immunogen E. coli-derived recombinant human IGFL-3 Thr25-Pro125 Accession # Q6UXB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388555
Test Specificity Detects human IGFL-3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IGF-I, rhIGF-II, rhIGFL-1, rhIGFL-4, and recombinant mouse IGFL-3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.