Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGFLR1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8296N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8296N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8296N100 Supplier R&D Systems Supplier No. FAB8296N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGFLR1/TMEM149 Monoclonal specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGFLR1/TMEM149
Applications Flow Cytometry
Classification Monoclonal
Clone 905338
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9H665
Gene Alias FLJ22573, TMEM149, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4
Gene Symbols IGFLR1
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IGFLR1 Met1-Pro163 Accession # Q9H665
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79713
Test Specificity Detects human IGFLR1in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.