Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $584.50
Specifications
Antigen | IgG |
---|---|
Clone | 1268C |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11012-SP
![]() |
R&D Systems
MAB11012SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11012-100
![]() |
R&D Systems
MAB11012100 |
100 μg |
Each for $584.50
|
|
|||||
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for ELISA.Specifications
IgG | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Immunoglobulin G, ImmunoglobulinG | |
Purified IgG from human serum | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IgG in direct ELISAs. As a pan-Human IgG-specific antibody, sandwich immunoassay specificity is determined by the capture antibody. |
1268C | |
ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
3500 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title