Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG, F(ab)2, Biotin, Isotype control, Novus Biologicals™
SDP

Catalog No. NBP196827 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196827 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196827 Supplier Novus Biologicals Supplier No. NBP196827
Add to Cart
Add to Cart

Human Antibody

IgG F(ab)2 specifically detects IgG F(ab)2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen IgG F(ab)2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Dot Blot
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Dot Blot
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Host Species Human
Purification Method Multi-step
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity No reaction was observed against anti-Human IgG F(c) or anti-Pepsin.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Human
Content And Storage Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.