Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG Fc APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB110A100
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.Specifications
IgG | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IGHG1 | |
Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region | |
100 Tests | |
Primary | |
Detects chimeric proteins containing a human IgG Fc region in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
97924 | |
Flow Cytometry 10 uL/10^6 cells | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3500 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction