Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG (gamma heavy chain) Rabbit anti-Human, Clone: RM116 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB14948
Description
Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain).
- High specificity
- High reactivity
Specifications
Human IgG (gamma heavy chain) | |
Monoclonal | |
Biotin | |
ELISA (0.05-0.2 ug/well for capture, 0.05-0.2 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) The optimal working dilution should be deter | |
P01857;P01859;P01860;P01861 | |
Rabbit | |
Protein A purification | |
RUO | |
3500/3501/3502/3503 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
RM116 | |
Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain). | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
IGHG1 | |
Native purified human IgG. | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction