Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG Heavy chain Monoclonal Antibody (54-76), Huabio

Catalog No. 502544502
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4502 Supplier HUABIO RESEARCH INC Supplier No. M08061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human IgG Heavy chain Monoclonal antibody specifically detects Human IgG Heavy chain in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen Human IgG Heavy chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone 54-76
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene IGHG4
Gene Accession No. P01861
Gene Alias IGHG1; IGHG4; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG
Host Species Mouse
Immunogen Native protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.