Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG Monoclonal Antibody (A5D12), Huabio

Catalog No. 502544513
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4513 Supplier HUABIO RESEARCH INC Supplier No. HA600076
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human IgG Monoclonal antibody specifically detects Human IgG in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen Human IgG
Applications ELISA, Western Blot
Classification Monoclonal
Clone A5D12
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Immunoglobulin G; ImmunoglobulinG
Host Species Mouse
Immunogen Full length native protein corresponding to Human IgG.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.