Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human IgG Monoclonal Antibody (KT131)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549527
Description
This product was used in an Indirect ELISA. This product does not cross-react with IgA, IgE and IgM.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifications
Human IgG | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
KT131 | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG | |
Recombinant Human IgG. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction