Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $514.50
Specifications
Antigen | IgG1 |
---|---|
Clone | 2347E |
Dilution | Western Blot 1 μg/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9930SP
![]() |
R&D Systems
MAB9930SP |
25 μg |
Each for $227.50
|
|
|||||
MAB9930100
![]() |
R&D Systems
MAB9930100 |
100 μg |
Each for $514.50
|
|
|||||
Description
IgG1 Monoclonal specifically detects IgG1 in Human samples. It is validated for Western Blot, ELISA.Specifications
IgG1 | |
Western Blot 1 μg/mL, ELISA | |
Monoclonal | |
Supernatant | |
Human | |
IgG1, Igh-4, RGD1359539, VH7183 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
2347E | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Full length Human IgG1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IgG1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title