Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.00
Specifications
Antigen | IgG1 |
---|---|
Clone | 984907 |
Dilution | Western Blot 2μg/mL, ELISA |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9894-SP
![]() |
R&D Systems
MAB9894SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9894-100
![]() |
R&D Systems
MAB9894100 |
100 μg |
Each for $561.00
|
|
|||||
Description
IgG1 Monoclonal specifically detects IgG1 in Human samples. It is validated for Western Blot, ELISA.Specifications
IgG1 | |
Western Blot 2μg/mL, ELISA | |
Monoclonal | |
Supernatant | |
Human | |
IgG1, Igh-4, RGD1359539, VH7183 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
984907 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Synthetic peptide containing human IgG 1 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IgG 1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title