Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $567.90
Specifications
| Antigen | IgG1 |
|---|---|
| Clone | 984932 |
| Dilution | Western Blot 2μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB98941-SP
![]() |
R&D Systems
MAB98941SP |
25 μg |
Each for $188.28
|
|
|||||
MAB98941-100
![]() |
R&D Systems
MAB98941100 |
100 μg |
Each for $567.90
|
|
|||||
Description
IgG1 Monoclonal specifically detects IgG1 in Human samples. It is validated for Western Blot.Specifications
| IgG1 | |
| Western Blot 2μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 984932 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Synthetic peptide containing human IgG 1 peptide | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human IgG 1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title