Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG1 Isotype Control, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. DDXCH01A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXCH01A647 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXCH01A647 Supplier Novus Biologicals Supplier No. DDXCH01A647100
Add to Cart
Add to Cart

Human Polyclonal Antibody

IgG1 Polyclonal specifically detects IgG1 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence.

Specifications

Antigen IgG1
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Alexa Fluor 647
Dilution Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500
Formulation PBS, 50% glycerol. with No Preservative
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Gene Symbols IGHG1
Host Species Human
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Isotype Control (Primary)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.