Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG1 Monoclonal Antibody (HP6069), Abbomax

Catalog No. 502204395
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4395 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4395 Supplier Abbomax Inc Supplier No. 605890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: plasma Cellular location: Secreted.

Human IgG1 Monoclonal antibody specifically detects Human IgG1 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Human IgG1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HP6069
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene IGHG1
Gene Accession No. P01857
Gene Alias IgG1; IgG3; Igh-4; Ighg; IGHG1; IGHG2; IGHG3; IGHG4; Immunoglobulin heavy chain (gamma polypeptide); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); RGD1359539; VH7183
Host Species Mouse
Immunogen Human immunoglobulin IgG1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.