Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | Human IgG2 |
---|---|
Clone | 2348B |
Dilution | Western Blot 2 μg;/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97941SP
![]() |
R&D Systems
MAB97941SP |
25 μg |
Each for $214.00
|
|
|||||
MB97941100
![]() |
R&D Systems
MAB97941100 |
100 μg |
Each for $526.00
|
|
|||||
Description
Human IgG2 Monoclonal specifically detects Human IgG2 in Human samples. It is validated for Western Blot, ELISA.Specifications
Human IgG2 | |
Western Blot 2 μg;/mL, ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Ig gamma-2 chain C region, IGHG2, immunoglobulin heavy constant gamma 2 (G2m marker) | |
Human IgG2 protein | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IgG2 in direct ELISAs. It does not detect human IgG1, IgG3 and IgG4 in direct ELISAs and Western Blots. |
2348B | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
3501.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title