Learn More
Invitrogen™ Human IgG3 Monoclonal Antibody (8D9)

Description
This product was used in both Indirect and Sandwich ELISA. This product does not cross-react with human IgG1, IgG2, IgG4, IgA, IgM and IgE.
Specifications
Specifications
| Antigen | Human IgG3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 8D9 |
| Concentration | 0.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene Alias | IgG3; IGHG3; immunoglobulin heavy constant gamma 3 (G3m marker) |
| Host Species | Mouse |
| Immunogen | Recombinant Human IgG3. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.