Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG4 (Fc) Rabbit anti-Human, Clone: RM217 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB14960
Description
Rabbit monoclonal antibody raised against synthetic peptide of human IgG4 (Fc).
- High specificity
- High reactivity
Specifications
Human IgG4 (Fc) | |
Monoclonal | |
Biotin | |
ELISA (0.05-0.2 ug/well for capture, 0.05-0.2 ug/mL for detection) Flow Cytometry The optimal working dilution should be determined by the end user. | |
P01861 | |
IGHG4 | |
A synthetic peptide corresponding to the Fc region of human IgG4. | |
50 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry | |
RM217 | |
Rabbit monoclonal antibody raised against synthetic peptide of human IgG4 (Fc). | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
MGC117419 | |
Rabbit | |
Protein A purification | |
RUO | |
3503 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction