Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgM Antibody (ICL-931), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142268AF750
Description
Human IgM Monoclonal antibody specifically detects Human IgM in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays, Dot BlotSpecifications
| Human IgM | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Human | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays (RIA) | |
| ICL-931 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Radioimmunoassay, Dot Blot | |
| AGM1, MU, VH | |
| Purified human IgM | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction