Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgM Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$212.05 - $567.90
Specifications
| Antigen | IgM |
|---|---|
| Clone | 2123C |
| Dilution | Western Blot 0.1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB94351SP
![]() |
R&D Systems
MAB94351SP |
25 μg |
Each for $212.05
|
|
|||||
MAB94351100
![]() |
R&D Systems
MAB94351100 |
100 μg |
Each for $567.90
|
|
|||||
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
| IgM | |
| Western Blot 0.1 ug/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Immunoglobulin M (IgM), mu chain from human plasma | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IgM in direct ELISAs and Western blots. |
| 2123C | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 3507 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title