Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HUMAN IgM Rabbit anti-Human, Clone: RM121, Abnova™

Catalog No. 89934332 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-332 Supplier Abnova Corporation Supplier No. MAB12812
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against native human IgM.

Specifications

Antigen HUMAN IgM
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RM121
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against native human IgM.
Dilution ELISA (0.05-0.2 ug/mL for detection; 0.05-0.2 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias DKFZp686I15196/DKFZp686I15212/FLJ00385/MGC104996/MGC52291/MU/VH
Gene Symbols IGHM
Host Species Rabbit
Immunogen Native purified IgM from human plasma.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3507
Test Specificity This antibody reacts to the heavy chain of human IgM. No cross reactivity with human IgG, IgA, IgD, or IgE.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.