Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGSF3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4788T100UG
This item is not returnable.
View return policy
Description
IGSF3 Monoclonal specifically detects IGSF3 in Human samples. It is validated for Flow Cytometry.Specifications
IGSF3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAI44133 | |
IGSF3 | |
Mouse myeloma cell line NS0-derived recombinant human IGSF3 Gln20-Ala1125 Accession # AAI44133 | |
RUO | |
3321 | |
Human | |
IgG1 |
Flow Cytometry | |
503621 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EWI-3, glu-Trp-Ile EWI motif-containing protein 3, immunoglobulin superfamily, member 3, member 3, V8 | |
Mouse | |
100 μg | |
Primary | |
Detects human IGSF3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IGSF4, 4B, 4C, 4D or 8 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction