Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGSF4C/SynCAM4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB41641
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB41641 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB41641 Supplier R&D Systems Supplier No. MAB41641
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IGSF4C/SynCAM4 Monoclonal specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen IGSF4C/SynCAM4
Applications Immunohistochemistry
Classification Monoclonal
Clone 404617
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NFZ8
Gene Alias CADM4, cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2, TSLL2SynCAM4
Gene Symbols CADM4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4 Gln25-Tyr323 Accession # Q8NFZ8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199731
Test Specificity Detects human IGSF4C/SynCAM4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IGSF4A, recombinant human (rh) IGSF3, rhIGSF4B, rhIGSF4D, or rhIGSF8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.