Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ikaros Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4984SP
This item is not returnable.
View return policy
Description
Ikaros/IKZF1 Polyclonal specifically detects Ikaros/IKZF1 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, CyTOF-ready.Specifications
Ikaros/IKZF1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DNA-binding protein Ikaros, hIk-1, IK1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, IKZF1, LyF-1, LYF1, LYF1PRO0758, PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1, ZNFN1A1CLL-associated antigen KW-6 | |
Goat | |
Affinity Purified | |
RUO | |
10320 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ChIP Assay, Flow Cytometry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
Q13422 | |
IKZF1 | |
E. coli-derived recombinant human lkaros Ser427-Ser519 Accession # Q13422 | |
25 μg | |
Primary | |
Detects human lkaros in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinat human (rh) ZIC-1, rhZNF-24, and rhZNF-206 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction