Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IkB-epsilon Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4300 Supplier R&D Systems Supplier No. AF4300
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IkB-epsilon Polyclonal specifically detects IkB-epsilon in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen IkB-epsilon
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. O00221
Gene Alias epsilon, ikappaBepsilon, I-kappa-B-epsilon, IkB-E, IKBENF-kappa-B inhibitor epsilon, ikB-epsilon, IkBepsilon, NF-kappa-BIE, NFKBIE, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene Symbols NFKBIE
Host Species Goat
Immunogen E. coli-derived recombinant human IkB-epsilon Ser141-Asp500 Accession # O00221
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4794
Test Specificity Detects human IkB-epsilon in direct ELISAs. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse IkB-epsilon is observed and less than 1% cross-reactivity with recombinant human IkB-beta is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.